Cables
170, InLab® cable S7-no connector, 5 m
Katalogové číslo 661-0269
Dodavatel: Mettler - Toledo
Přejít na:
Product Details & Documents
These electrode cables are designed to connect Mettler sensors to almost any meter.
Documents
Mettler InLab® Sensors
Produktová literatura
Specifikace
- Popis:InLab® cable S7-no connector
- Délka:5 m
- Sekvence:170
- Cat. No.:661-0269