Orders LinkContactUsLinkComponent
Cables
Cables
165, InLab® cable S7-4mm, 1,2 m
Catalog # 661-0264
Supplier:   Mettler - Toledo
Cables
Catalog # 661-0264
Supplier:  Mettler - Toledo
Cables
Cables
165, InLab® cable S7-4mm, 1,2 m
Price
2 010,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
These electrode cables are designed to connect Mettler sensors to almost any meter.
Documents
Specifications
  • Popis:
    InLab® cable S7-4mm
  • Délka:
    1,2 m
  • Sekvence:
    165