Orders LinkContactUsLinkComponent
Cables
158, InLab® cable S7-BNC, 1,2 m
Catalog # 661-0257
Supplier:   Mettler - Toledo
Cables
Catalog # 661-0257
Supplier:  Mettler - Toledo
Cables
Cables
158, InLab® cable S7-BNC, 1,2 m
Price
1 810,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
These electrode cables are designed to connect Mettler sensors to almost any meter.
Specifications
  • Popis:
    InLab® cable S7-BNC
  • Délka:
    1,2 m
  • Sekvence:
    158