Orders LinkContactUsLinkComponent
Cables
Cables
157, InLab® cable MultiPin-BNC/RCA (IP67), 1,8 m
Catalog # 661-0256
Supplier:   Mettler - Toledo
Cables
Catalog # 661-0256
Supplier:  Mettler - Toledo
Cables
Cables
157, InLab® cable MultiPin-BNC/RCA (IP67), 1,8 m
Price
4 490,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
These electrode cables are designed to connect Mettler sensors to almost any meter.
Documents
Specifications
  • Popis:
    InLab® cable MultiPin-BNC/RCA (IP67)
  • Délka:
    1,8 m
  • Sekvence:
    157