Orders LinkContactUsLinkComponent
Embedding cassettes with disposable lid
02, Žlutá
Katalogové číslo KLIN110-02S
Dodavatel:   KLINIPATH
Embedding cassettes with disposable lid
Katalogové číslo KLIN110-02S
Dodavatel:  KLINIPATH
Embedding cassettes with disposable lid
Embedding cassettes with disposable lid
02, Žlutá
Přejít na:
Price
4 530,00 Kč
Přihlásit se pro dostupnost
1 * 1.600 KS
Product Details & Documents
Embedded cassettes with hinged lid.
  • Cassettes maintain their stability, rigidness and shape during processing of tissues
 : Available in different colours.
 : Hoppers contain 80 cassettes which are specially packed for Thermo Fisher® cassette printers. Taped stacks contain 40 cassettes which are specially packed for Sakura® / Leica® cassette printers. Lids are packed separately.
Specifikace
  • Barva:
    Žlutá
  • Sekvence:
    02