ION STANDARD NA 10 MG/L 1 * 500 mL
Catalog # METT30090858
Supplier: Mettler - Toledo
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Conductivity standards
Supplier: Mettler - Toledo
Chlorid draselný 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
VWR® AVS TITRINORM, pufrovací roztoky, 20 °C
Supplier: VWR Chemicals
Roztoky pufrů v sáčcích, Certipur®, Supelco®
Supplier: Merck
Roztoky standardní pro měření vodivosti
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
pH buffers, ready to use
Supplier: WTW
Visual comparator test kits, dissolved oxygen by rhodazine method, CHEMets
Supplier: CHEMetrics