Seed testing papers, Munktell
01, 37/N, 70 mm
BINZ233208
: Ahlstrom
Seed Testing Papers are widely used by seed testing institutes and others for germination capacity testing and for health tests e.g. Jacobsen (Copenhagen tank), on top of paper and osmosis test.
- Grade/stupeň/jakost:37/N
- Sekvence:01
- Velikost:70 mm
- Hmotnost:135 g/m²
- Cat. No.:BINZ233208
- Třída:37/N
- Kapilární jev:-
- Obsah popela:-