Atazanavir, Reference Standards
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Atazanavir System Suitability Mixture: A mixture of atazanavir sulfate, atazanavir R, S, S, S,-diastereomer and atazanavir S,R,S,S diastereomer
Certifications: US Pharmacopea Reference Standard
Specifikace
Specifikace
- Typ balení:Vial
- Bal.:200 mg
- Standard:USP reference standard
- Popis:Atazanavir Sulfate
- Popis:Atazanavir Sulfate
- Cat. No.:1044334.
- Čí. CAS:229975-97-7
- Vzorec:C₃₈H₅₂N₆O₇, H₂O₄S
Specifikace
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.