Orders LinkContactUsLinkComponent
Permeability cups
01, Permeability cup with a surface area of 10 cm²
Katalogové číslo SIMEVF2200
Dodavatel:   Industrial Physics
Permeability cups
Katalogové číslo SIMEVF2200
Dodavatel:  Industrial Physics
Permeability cups
Permeability cups
01, Permeability cup with a surface area of 10 cm²
Přejít na:
Price
6 600,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Permeability cups are designed to determine the water vapour transmission of paints, varnishes, coatings and related products. Permeability cup is suitable for testing both self-supporting coating and non-self-supporting coatings.
  • Anodised aluminium for corrosion resistance and colouring
  • Seal ring to prevent turning when closing the cover
  • Any deformation to the cup may cause deviation in the result
The wet cup method is the reference for determining water vapour transmission.
Certifications: ISO 7783 (supersedes NF T30-018), ASTM D1653, ASTM E95.
Delivery information: Includes a cup, a seal ring and cover ring.
Specifikace
  • Popis:
    Permeability cup with a surface area of 10 cm²
  • Sekvence:
    01