Orders LinkContactUsLinkComponent
eXtreme/FV® Filter Vials
eXtreme/FV® Filter Vials
10, Nylon, Tiered filtration, eXtreme/FV® Filter vial, 0,45 µm, Růžová
Katalogové číslo THMS85539-500
Dodavatel:   THOMSON INSTRUMENT
eXtreme/FV® Filter Vials
Katalogové číslo THMS85539-500
Dodavatel:  THOMSON INSTRUMENT
eXtreme/FV® Filter Vials
eXtreme/FV® Filter Vials
10, Nylon, Tiered filtration, eXtreme/FV® Filter vial, 0,45 µm, Růžová
Přejít na:
Price
54 610,00 Kč
Přihlásit se pro dostupnost
1 * 500 KS
Product Details & Documents
Thomson's filtration products are engineered and optimised to increase the efficiency of sample preparation without compromising the results.
Thomson eXtreme/FV® Filter vials offer tiered filtration, and are designed to speed up sample prep and analysis on particulate laden samples. Save on costly repairs and service issues while eliminating multiple syringe filters, syringes, HPLC vials, and HPLC caps. The eXtreme/FV® Filter Vials are easy to press down with hands or toggle press.
These filter vials are manufactured with attention to detail and high-grade quality to ensure performance at optimal levels. Filter vials find a wide variety of applications ranging from life sciences, drug discovery, monitoring, and analysis. They are universal for all different types of autosamplers, including UHPLC.
Specifikace
  • Membrána:
    Nylon, Tiered filtration
  • Velikost pórů:
    0,45 µm
  • Barva:
    Růžová
  • Popis:
    eXtreme/FV® Filter vial
  • Sekvence:
    10