Orders LinkContactUsLinkComponent
Atazanavir, Reference Standards
Atazanavir, Reference Standards
Atazanavir System Suitability Mixture, USP reference standard, Vial, 25 mg
Katalogové číslo 1044345.
Dodavatel:   USP
Atazanavir, Reference Standards
Katalogové číslo 1044345.
Dodavatel:  USP
Číslo CAS:  
Atazanavir, Reference Standards
Atazanavir System Suitability Mixture, USP reference standard, Vial, 25 mg
Přejít na:
Číslo CAS:  
Price
30 770,00 Kč
Přihlásit se pro dostupnost
1 * 25 mg
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Atazanavir System Suitability Mixture: A mixture of atazanavir sulfate, atazanavir R, S, S, S,-diastereomer and atazanavir S,R,S,S diastereomer
Certifications: US Pharmacopea Reference Standard
Specifikace
Specifikace
  • Typ balení:
    Vial
  • Bal.:
    25 mg
  • Standard:
    USP reference standard
  • Popis:
    Atazanavir System Suitability Mixture