methylcyklohexan 99% for spectroscopy
5096, Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
methylcyklohexan 99% for spectroscopy
Catalog # ACRO433770025
Supplier: Thermo Fisher Scientific
CAS Number: 108-87-2
Jump to:
Product Details & Documents
- CAS Number:108-87-2
- Formula:C₆H₁₁CH₃
- MDL Number:MFCD00001497
- Molecular Weight:98.19
- Boiling Pt:101 °C (760 mmHg)
- Melting Pt:–126,3 °C
- Density:0,77 g/cm³
- Flash Pt:–3 °C
- Storage Temperature:Ambient
- UN:2296
- ADR:3,II
- :Density: 0,7693 g/cm³ (20 °C)
Documents
ACRO43377
SDS
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Water:≤0.5 %
- GC:≥98.5 %
- UV:at 230 nm A: ≤0.15
- Color scale:≤5 APHA
- Residue after evaporation:≤0.001 %
Specifications
- Typ balení:Glass bottle
- Bal.:2,5 L
- Sekvence:5096



