Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Horizontal wafer shippers, contactless, 200 mm SmartStack®
Horizontal wafer shippers, contactless, 200 mm SmartStack®
04, Contactless horizontal wafer shipper kit
Katalogové číslo ENTEHWS200C-1PK-KI
Dodavatel:   ENTEGRIS
Horizontal wafer shippers, contactless, 200 mm SmartStack®
Katalogové číslo ENTEHWS200C-1PK-KI
Dodavatel:  ENTEGRIS
Horizontal wafer shippers, contactless, 200 mm SmartStack®
04, Contactless horizontal wafer shipper kit
Přejít na:
Price
Poptat cenu
Přihlásit se pro dostupnost
1 * 2 KS
Product Details & Documents
This SmartStack® contactless horizontal wafer shippers safely protect and transport bumped wafers in full, thin or ultrathin thickness.
  • Transports up to 25 wafers of varying thickness
  • Stackable configuration increases shipping density and is automation compatible
  • Used for thin, 3D, lensed or bumped wafers
Perimeter support ring minimizes the potential for surface area damage and eliminates the likelihood of scuffed, marred or bump damaged wafers. Rugged, engineering-grade polymer construction eliminates the need for TYVEK® separators and pink foam cushions.
Ordering information: Shipper contains contactless horizontal wafer shipper and 26 rings. Shipper kit contains contactless horizontal wafer shipper, 26 rings, corrugated shipping box, outer packing cushions and CMB moisture barrier bag.
Specifikace
  • Popis:
    Contactless horizontal wafer shipper kit
  • Sekvence:
    04
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.