Horizontal wafer shippers, contactless, 200 mm SmartStack®
Product Details & Documents
This SmartStack® contactless horizontal wafer shippers safely protect and transport bumped wafers in full, thin or ultrathin thickness.
- Transports up to 25 wafers of varying thickness
- Stackable configuration increases shipping density and is automation compatible
- Used for thin, 3D, lensed or bumped wafers
Perimeter support ring minimizes the potential for surface area damage and eliminates the likelihood of scuffed, marred or bump damaged wafers. Rugged, engineering-grade polymer construction eliminates the need for TYVEK® separators and pink foam cushions.
Ordering information: Shipper contains contactless horizontal wafer shipper and 26 rings. Shipper kit contains contactless horizontal wafer shipper, 26 rings, corrugated shipping box, outer packing cushions and CMB moisture barrier bag.
Specifikace
- Popis:Contactless horizontal wafer shipper kit
- Sekvence:04