Orders LinkContactUsLinkComponent
Lock boards
Lock boards
01, Lockout station for 5 locks, Žlutá, 393×292 mm
Katalogové číslo 238-0191
Dodavatel:   Brady
Lock boards
Katalogové číslo 238-0191
Dodavatel:  Brady
Lock boards
Lock boards
01, Lockout station for 5 locks, Žlutá, 393×292 mm
Přejít na:
Price
6 490,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
These acrylic lockout stations are suitable for storing tags, padlocks and hasps for either 5, 10, 26 or 36 people.
  • Highly visible thanks to yellow colour
  • For wall mounting
Please note: Tags, padlocks and hasps are not included.
Specifikace
  • Type:
    Lockout station for 5 locks
  • Barva:
    Žlutá
  • Sekvence:
    01
  • Š×H×V:
    393×292 mm