Orders LinkContactUsLinkComponent
VWR® Acetate Buffer USP Test Solution (TS)
VWR® Acetate Buffer USP Test Solution (TS)
02, Acetate buffer TS, Plastic bottle, 500 mL
Katalogové číslo 85323.260
Dodavatel:   VWR Chemicals
VWR® Acetate Buffer USP Test Solution (TS)
Katalogové číslo 85323.260
Dodavatel:  VWR Chemicals
Číslo CAS:  
VWR® Acetate Buffer USP Test Solution (TS)
VWR® Acetate Buffer USP Test Solution (TS)
02, Acetate buffer TS, Plastic bottle, 500 mL
Přejít na:
Číslo CAS:  
Price
7 160,00 Kč
Přihlásit se pro dostupnost
1 * 500 mL
Product Details & Documents
Ready to use test solutions (TS).
  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available
Specifikace
Specifikace
  • Typ balení:
    Plastic bottle
  • Bal.:
    500 mL
  • Popis:
    Acetate buffer TS
  • Sekvence:
    02