Orders LinkContactUsLinkComponent
BD® Blunt Needles
BD® Blunt Needles
03, 18 G, 40 mm, Blunt bevel, Sterile, Červený
Katalogové číslo BDAM303129
Dodavatel:   BD Medical
BD® Blunt Needles
Katalogové číslo BDAM303129
Dodavatel:  BD Medical
BD® Blunt Needles
03, 18 G, 40 mm, Blunt bevel, Sterile, Červený
Přejít na:
Price
765,00 Kč
Přihlásit se pro dostupnost
1 * 100 KS
Product Details & Documents
BD blunt fill and blunt filter needles are specifically designed to reduce the risk of needlestick injuries during medication preparation.
  • The blunt fill needle is designed for vial access and available with a BD Luer-Lok™ connection
  • The blunt filter is designed for safe ampoule medication preparation
  • BD blunt fill needles have female luer fittings, which mate to male luer fittings
  • BD blunt filter needle has a 5-micron filter for particles such as glass and plastic fragments, for safer medication delivery
  • Natural rubber latex is not part of the material formulation
  • Sterile, singe use
 : Note: The BD blunt fill and filter needles are not for skin injection.
Specifikace
  • Barevný kód:
    Červený
  • Rozchod:
    18 G
  • Type:
    Sterile
  • Typ špičky:
    Blunt bevel
  • Délka:
    40 mm
  • Sekvence:
    03