Orders LinkContactUsLinkComponent
Wall cleaning tool
Wall cleaning tool
01, Wall cleaning tool stainless steel: For wall and ceiling cleaning, with handle pick-up made of stainless steel, - mm
Katalogové číslo PPSA3580033
Wall cleaning tool
Katalogové číslo PPSA3580033
Wall cleaning tool
Wall cleaning tool
01, Wall cleaning tool stainless steel: For wall and ceiling cleaning, with handle pick-up made of stainless steel, - mm
Přejít na:
Price
2 470,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Wall cleaning tool for wall and ceiling cleaning in hygienically sensitve areas.
  • Frame completely made of stainless steel
  • Mops are 100% polyester microfiber filament yarn
  • Autoclavable, disinfectant proof
  • Mops have high-quality stainless steel push buttons for fitting with frame
This mop frame with locking screw is suitable with mop covers Micro Sicuro CR/A and Mop CR for wall cleaning tool.
 : Mops are prewashed and single double packaged.
Documents
Specifikace
  • Popis:
    Wall cleaning tool stainless steel: For wall and ceiling cleaning, with handle pick-up made of stainless steel
  • D׊:
    - mm
  • Sekvence:
    01