Orders LinkContactUsLinkComponent
Precision butyrometer, without stopper
Precision butyrometer, without stopper
03, Butyrometer, precision, 0 - 4 % : 0,05, with conformity certificate, without stopper
Katalogové číslo GERB3150-C-O
Dodavatel:   GERBER FUNKE
Precision butyrometer, without stopper
Katalogové číslo GERB3150-C-O
Dodavatel:  GERBER FUNKE
Precision butyrometer, without stopper
03, Butyrometer, precision, 0 - 4 % : 0,05, with conformity certificate, without stopper
Price
636,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
For drinking vat milk, frosted rear scale wall, error tolerance 0,025%.
Specifikace
  • Popis:
    Butyrometer, precision, 0 - 4 % : 0,05, with conformity certificate, without stopper
  • Sekvence:
    03
Accessories
Informace o produktu
  • Katalogové číslo
    Dostupnost
    Price
  • Stopper, rubber, conical, for all measuring method butyrometers
    Stopper, rubber, conical, for all measuring method butyrometers
    614-2028
    Přihlásit se pro dostupnost
    1 * 100 KS - 1 170,00 Kč