Pipette tips, epT.I.P.S.® Motion
21, Standard, 1 - 50 µl, -, 10 racks, 96 each
Katalogové číslo EPPE0030014.405
Dodavatel: EPPENDORF
Přejít na:
Product Details & Documents
These pipette tips are specially designed for use on the epMotion 5070 and 5075 Liquid Handling Workstation. They were developed in torsion-resistant, contamination-free racks.
- High-precision manufacturing guarantees a perfect fit and extremely accurate dispensing behavior
- Tips are optimised for simple automatic ejection in the Liquid Handling Workstation collection vessel
- Tips can also be integrated into the workstation as a refilling system
- Refilling tips are delivered in a PET thermoformed tray with sealed lid
- Replacing the disposable box with an autoclavable aluminum adapter can mean a waste savings of up to 40 percent
The epT.I.P.S. Motion Rack is made up of two components for flexible use in the automated epMotion Liquid Handling Workstation: a colour-coded tray with the epT.I.P.S. filter tips and a disposable box for storing the tray.
Balení: Packaged in contamination-free racks or in reload trays.
Specifikace
- Balené:10 racks, 96 each
- Sterilní:-
- Objem:1 - 50 µl
- Sekvence:21
- Druh špičky:Standard