Orders LinkContactUsLinkComponent
Volumetric flasks, class A
Volumetric flasks, class A
28, 1000 ml, 24/20, 0,40 ± ml
Catalog # POULRB258/K/2
Supplier:   Poulten & Graf
Volumetric flasks, class A
Catalog # POULRB258/K/2
Supplier:  Poulten & Graf
Volumetric flasks, class A
Volumetric flasks, class A
28, 1000 ml, 24/20, 0,40 ± ml
Price
1 850,00 Kč
Log in for availability
1 * 2 KS
Product Details & Documents
Borosilicate glass, with PP NS-stopper.
Individually numbered.
Certifications: DIN/BS/ISO 1042
Documents
Specifications
  • NZ:
    24/20
  • Kapacita:
    1000 ml
  • Sekvence:
    28
  • Tolerance:
    0,40 ± ml