Volumetric flasks, class A
19, 10 ml, 10/19, 0,025 ± ml
Catalog # POULRB258/B/5
Supplier: Poulten & Graf
Přejít na:
Product Details & Documents
Borosilicate glass, with PP NS-stopper.
Individually numbered.
Certifications: DIN/BS/ISO 1042
Documents
POULRB258^WAC^J^2_TDS_EN
Datasheet
Specifikace
- NZ:10/19
- Kapacita:10 ml
- Sekvence:19
- Tolerance:0,025 ± ml