Přeskočit na Záhlaví
Přeskočit na Hlavní obsah
Přeskočit na Patička
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Copper(II) sulfate pentahydrate powder suitable for use as excipient EMPROVE® exp Ph Eur,BP,USP 1 * 0,75 kg
Catalog # 1.02788.0750
Supplier:
MERCK PRODUCTION CHEMICALS
SDS
Certificates
Expand image
Copper(II) sulfate pentahydrate powder suitable for use as excipient EMPROVE® exp Ph Eur,BP,USP 1 * 0,75 kg
Catalog # 1.02788.0750
Supplier:
MERCK PRODUCTION CHEMICALS
SDS
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Product specification
MERP1.02788.0750_TDS_EN
SDS
msds_102788
Documentation
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Chlorid draselný, EMSURE® p.a., Supelco®
Supplier: Merck
Voda, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Jodid draselný ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Dihydrogenfosforečnan draselný, EMSURE® ISO p.a., Supelco®
Supplier: Merck
Acetonitril ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
Supplier: Merck
Síran sodný, anhydrous, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
Supplier: Merck
Serological pipettes, Stripette™
Supplier: Corning
Chlorid sodný, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
Supplier: Merck
Jodid draselný ≥99.5%, EMSURE® ISO, Reag. Ph. Eur. p.a., Supelco®
Supplier: Merck
Chlorid sodný regeneration salt, TECHNICAL for water softening
Supplier: VWR Chemicals
Chlorid sodný 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitril, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® USP, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Síran železnatý heptahydrát 99.5-105.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals