[EN]ACE 3 C4 NANO COLUMN 150 X 0.075 1 * 1 KS
HICHACE1131500075
: Avantor
Frequently Bought Together
Voda, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Aceton ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Ubrousky čisticí
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Rukavice, nitrilové, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Sérologická pipeta, standardní
: VWR Collection
VWR® NITRILE LIGHT, Rukavice, nitrilové
: VWR Collection
VWR®, Pipety Pasteurovy, High performance
: VWR Collection