Orders LinkContactUsLinkComponent
Connectors, PP
Connectors, PP
05, Connector, straight, male luer lock/tubing nipple, 4,8 mm
Catalog # INFIPP-LMS48
Supplier:   PIEPER FILTER
Connectors, PP
Catalog # INFIPP-LMS48
Supplier:  PIEPER FILTER
Connectors, PP
Connectors, PP
05, Connector, straight, male luer lock/tubing nipple, 4,8 mm
Price
734,00 Kč
Log in for availability
1 * 10 KS
Product Details & Documents
Connectors, made of PP, natural.
  • Good chemical resistance
  • Good temperature resistance
Specifications
  • Hadice int.Ø:
    4,8 mm
  • Popis:
    Connector, straight, male luer lock/tubing nipple
  • Sekvence:
    05