Orders LinkContactUsLinkComponent
Modular storage systems, KP
Modular storage systems, KP
KP-BLOCK, modular and stackable block storage metal cabinet with 13 drawers, red - capacity: 700 blocks
Katalogové číslo 720-2810
Dodavatel:   KLINIPATH
Modular storage systems, KP
Katalogové číslo 720-2810
Dodavatel:  KLINIPATH
Modular storage systems, KP
Modular storage systems, KP
KP-BLOCK, modular and stackable block storage metal cabinet with 13 drawers, red - capacity: 700 blocks
Price
Poptat cenu
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Stackable, universal storage system for preparations and blocks. A complete system with growth potential. A modular system able to transform local storage in a laboratory into a long-term storage system. A growth system with easy exchangeable and removable plastic trays.
  • Lower gliding resistance, no rusting because of paint damage
Specifikace
  • Popis:
    KP-BLOCK, modular and stackable block storage metal cabinet with 13 drawers, red - capacity: 700 blocks
Accessories
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Top for modular systems, KP Glass and KP Block
    Top for modular systems, KP Glass and KP Block
    720-2813
    02
    Přihlásit se pro dostupnost
    1 * 1 KS - 186,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Base for modular systems, KP Glass and KP Block
    Base for modular systems, KP Glass and KP Block
    720-2812
    01
    Přihlásit se pro dostupnost
    1 * 1 KS - 1 240,00 Kč