Modular storage systems, KP
KP-BLOCK, modular and stackable block storage metal cabinet with 13 drawers, blue - capacity: 700 blocks
Katalogové číslo 720-2809
Dodavatel: KLINIPATH
Přejít na:
Product Details & Documents
Stackable, universal storage system for preparations and blocks. A complete system with growth potential. A modular system able to transform local storage in a laboratory into a long-term storage system. A growth system with easy exchangeable and removable plastic trays.
- Lower gliding resistance, no rusting because of paint damage
Specifikace
- Popis:KP-BLOCK, modular and stackable block storage metal cabinet with 13 drawers, blue - capacity: 700 blocks
Accessories
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Top for modular systems, KP Glass and KP BlockTop for modular systems, KP Glass and KP Block720-281302Přihlásit se pro dostupnost1 * 1 KS - 186,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Base for modular systems, KP Glass and KP BlockBase for modular systems, KP Glass and KP Block720-281201Přihlásit se pro dostupnost1 * 1 KS - 1 240,00 Kč