Orders LinkContactUsLinkComponent
Wipe down table
Wipe down table
Wipe down table, with waste disposal hole on top and a cabinet under the table with removable plastic bin, 900×600×900 mm
Catalog # 132-1091
Supplier:   Palbam
Wipe down table
Catalog # 132-1091
Supplier:  Palbam
Wipe down table
Wipe down table
Wipe down table, with waste disposal hole on top and a cabinet under the table with removable plastic bin, 900×600×900 mm
Price
105 200,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
The wipe down table is designed to support the wipe down procedure. Includes a hole in the table top for easy waste disposal.
The cabinet under the table and under the waste disposal hole has a removable plastic bin included inside.
In stainless steel 304 with electropolished finish.
Specifications
  • Popis:
    Wipe down table, with waste disposal hole on top and a cabinet under the table with removable plastic bin
  • Š×H×V:
    900×600×900 mm