SODIUM HYDROXIDE 0.2 MOL/L (0.2N) STANDA 1 * 1.000 mL
Katalogové číslo APPC182971.1211
Dodavatel: PanReac AppliChem
SODIUM HYDROXIDE 0.2 MOL/L (0.2N) STANDA 1 * 1.000 mL
Katalogové číslo APPC182971.1211
Dodavatel: PanReac AppliChem
Frequently Bought Together
n-Hexan, EMSURE® ACS, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck
Diethylenglykolmonobutylether, Sigma-Aldrich®
Dodavatel: Merck
n-Hexan, dehydrated (max. 0.01% H₂O) ≥95%, GPR RECTAPUR® for synthesis
Dodavatel: VWR Chemicals
n-Pentan ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Dodavatel: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
1-Propanol, EMSURE® ACS, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck
1,2-Dichlorethan ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Tetrahydrofuran, EMPLURA®, extra pure, Supelco®
Dodavatel: Merck
Akrylonitril 99+%
Dodavatel: Thermo Fisher Scientific
Dimethylsulfoxid ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Dodavatel: Honeywell Chemicals
Glycerin (synthetic) 99+%, pure
Dodavatel: Thermo Fisher Scientific
1-methyl-2-pyrolidon 99+% ACS
Dodavatel: Thermo Fisher Scientific
Xylen (směs izomerů) for histology
Dodavatel: Honeywell Chemicals
Pyridin, extra dry over Molecular Sieve 99.5%, AcroSeal®
Dodavatel: Thermo Fisher Scientific
jodmetan ≥99% stabilizovaný
Dodavatel: Thermo Fisher Scientific
Glycerin, anhydrous ≥99.5% (by GC), Puriss. p.a. ACS, distilled, Riedel-de Haën™
Dodavatel: Honeywell Chemicals
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Dodavatel: VWR Chemicals