Orders LinkContactUsLinkComponent
Multi-channel pipettes, mechanical, variable volume, WITOPET-Multi
Multi-channel pipettes, mechanical, variable volume, WITOPET-Multi
01, ME 10, 0,5 - 10 µl, 0,1 µl, ≤ ±1,5 - ±4,0 %, ≤ ±1,5 - ±4,0 %
Katalogové číslo WITG5.403.101
Dodavatel:   witeg Labortechnik
Multi-channel pipettes, mechanical, variable volume, WITOPET-Multi
Katalogové číslo WITG5.403.101
Dodavatel:  witeg Labortechnik
Multi-channel pipettes, mechanical, variable volume, WITOPET-Multi
Multi-channel pipettes, mechanical, variable volume, WITOPET-Multi
01, ME 10, 0,5 - 10 µl, 0,1 µl, ≤ ±1,5 - ±4,0 %, ≤ ±1,5 - ±4,0 %
Přejít na:
Price
11 530,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
These multi-channel pipettes are designed for comfort over long term repetitive pipetting. 8 or 12 identical volumes for titration plates.
  • Highest possible accuracy and precision
  • Easy readable digital volume display
  • Volume ranges colour coded on the push button
  • Volumes steplessly adjustable
  • Tip ejector can be blocked to avoid mis-application when using the instrument over large series
 : Each pipette is individually tested and delivered with a test certificate.
Specifikace
  • Objem:
    0,5 - 10 µl
  • Popis:
    ME 10
  • Sekvence:
    01
  • Increment:
    0,1 µl
  • Přesnost:
    ≤ ±1,5 - ±4,0 %
  • Správnost:
    ≤ ±1,5 - ±4,0 %