Orders LinkContactUsLinkComponent
Conductivity standards
08, Conductivity standard solution, 1413 µS/cm, 250 mL
  HACHLZW9710.99
 :   Hach
Conductivity standards
  HACHLZW9710.99
 :  Hach
 :  
Conductivity standards
Conductivity standards
08, Conductivity standard solution, 1413 µS/cm, 250 mL
 :  
782,00 Kč
1 * 250 mL
The sodium chloride standard solutions are used for calibrating conductivity meters and also used for accuracy checks.
  • Bal.:
    250 mL
  • Popis:
    Conductivity standard solution
  • Sekvence:
    08
  • Vodivost:
    1413 µS/cm