Temperature indicating kit
03, Thermax chill checker, type 8, +20 °C
Katalogové číslo KLEI1216168
Dodavatel: KLEINFELD
Přejít na:
Product Details & Documents
These Thermax chill checker include eight different measurement points over a temperature range from −17 to +20 °C. Activation takes place after cooling by pressing in the center. When the temperature is exceeded, it changes color purple or blue.
Specifikace
- Max. tepl.:+20 °C
- Popis:Thermax chill checker, type 8
- Sekvence:03