Orders LinkContactUsLinkComponent
Corning® Lymphocyte Separation Medium, Corning
Corning® Lymphocyte Separation Medium, Corning
01, LSM (Lymphocyte Separation Medium), 1×100 ml
Katalogové číslo MDTC25-072-CI
Dodavatel:   Corning
Corning® Lymphocyte Separation Medium, Corning
Katalogové číslo MDTC25-072-CI
Dodavatel:  Corning
Corning® Lymphocyte Separation Medium, Corning
Corning® Lymphocyte Separation Medium, Corning
01, LSM (Lymphocyte Separation Medium), 1×100 ml
Přejít na:
Price
1 320,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Lymphocyte Separation Medium (LSM) was originally designed for the in vitro isolation of lymphocytes from diluted whole blood
It is a sterile, iso-osmotic polysucrose and diatrizoate solution with low viscosity and a density of 1.077-1.080 g/mL at 20ºC.
Specifikace
  • Popis:
    LSM (Lymphocyte Separation Medium), 1×100 ml
  • Sekvence:
    01