Connectors, PP
17, Connector, straight, female luer lug/female luer lug, —
Catalog # INFIPP-LFL/LFL
Supplier: PIEPER FILTER
Jump to:
Product Details & Documents
Connectors, made of PP, natural.
- Good chemical resistance
- Good temperature resistance
Specifications
- Hadice int.Ø:—
- Popis:Connector, straight, female luer lug/female luer lug
- Sekvence:17