Orders LinkContactUsLinkComponent
Safety spectacles, Tracker
Safety spectacles, Tracker
04, Anti-scratch, Černá, Welding shade 5 PC
Catalog # BOLETRACWPCC5
Supplier:   Bollé Safety
Safety spectacles, Tracker
Catalog # BOLETRACWPCC5
Supplier:  Bollé Safety
Safety spectacles, Tracker
Safety spectacles, Tracker
04, Anti-scratch, Černá, Welding shade 5 PC
Price
538,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
These sporty design safety spectacles feature a durable PC frame, a smoke PC dual lens, straight temples with upper and lower protection and an adjustable headband.
  • Interchangeable foam reinforcement with indirect ventilation
  • Adjustable head strap and rubber arm tips provide a secure fit and greater protection
  • ALS anti-fog/anti-scratch coating on both sides of clear and smoke lenses
  • Rubberised eye seal provides exceptional protection against dust and other particles
Weight: 52 g
EN 166
Certifications: EN 166
Delivery information: Supplied with microfibre pouch.
Specifications
  • Type:
    Anti-scratch
  • Barva:
    Černá
  • Sekvence:
    04
  • Čočky/objektiv:
    Welding shade 5 PC