Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PARAFORMALDEHYDE 4% 1 * 500 mL
Catalog # BSBTAR1068
Supplier:
Boster Bio
Expand image
PARAFORMALDEHYDE 4% 1 * 500 mL
Catalog # BSBTAR1068
Supplier:
Boster Bio
Related Information
Documentation
About Boster
Datasheet
AR1068
Additional Product Details
Literature reference - PubMed ID 21289500
Literature reference - PubMed ID 21505994
Literature reference - PubMed ID 22179675
Literature reference - PubMed ID 22710477
Literature reference - PubMed ID 24681123
More
Documentation
Frequently Bought Together
Ethylacetát, EMSURE® ACS, ISO, Reag. Ph. Eur. 1035300 p.a., Supelco®
Supplier: Merck
Methanol ≥99.9% (by GC), LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
Supplier: Merck
Diethylether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Voda USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Methanol, LiChrosolv®, hypergrade for LC-MS, for HPLC, Supelco®
Supplier: Merck
2-Propanol, LiChrosolv®, gradient grade for liquid chromatography, Supelco®
Supplier: Merck
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Aceton ≥99.9%, PESTINORM® Supra Trace for organic trace analysis
Supplier: VWR Chemicals
Toluen, anhydrous (max. 0.005% H₂O) ≥99.8%, AnalaR NORMAPUR® for synthesis
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Tetrahydrofuran stabilizovaný, EMSURE® ACS, Reag. Ph. Eur. p.a., Supelco®
Supplier: Merck
Voda, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Xylen (směs izomerů) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
1-Propanol ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals