Orders LinkContactUsLinkComponent
Test kits, drop count tests
Test kits, drop count tests
10, Formaldehyde, model FM-1, 0,5 - 10%, 1 KIT
Katalogové číslo HACH2183100
Dodavatel:   Hach
Test kits, drop count tests
Katalogové číslo HACH2183100
Dodavatel:  Hach
Číslo CAS:  
Test kits, drop count tests
Test kits, drop count tests
10, Formaldehyde, model FM-1, 0,5 - 10%, 1 KIT
Přejít na:
Číslo CAS:  
Price
5 710,00 Kč
Přihlásit se pro dostupnost
1 * 1 KIT
Product Details & Documents
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
  • Portable - ideal for use in field or in lab
  • Used as a quick reference or for confirmatory test with colour indicator
  • Reagents supplied with the kit
Upozornení: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifikace
Specifikace
  • Bal.:
    1 KIT
  • Popis:
    Formaldehyde, model FM-1
  • Sekvence:
    10
  • Rozsah:
    0,5 - 10%