Orders LinkContactUsLinkComponent
Dropping pipettes
Dropping pipettes
02, Dropping pipettes with silicone teat, for 100 ml bottles, joint size 14/23
Katalogové číslo BIBBSBD100
Dodavatel:   SciLabware
Dropping pipettes
Katalogové číslo BIBBSBD100
Dodavatel:  SciLabware
Dropping pipettes
Dropping pipettes
02, Dropping pipettes with silicone teat, for 100 ml bottles, joint size 14/23
Přejít na:
Price
2 520,00 Kč
Přihlásit se pro dostupnost
1 * 5 KS
Product Details & Documents
Soda-lime glass.
  • With integral ground glass cone on pipette
  • Supplied with rubber teats
  • Bottles available separately
Specifikace
  • Popis:
    Dropping pipettes with silicone teat, for 100 ml bottles, joint size 14/23
  • Sekvence:
    02