Disposable filters, Optimizer®
07, D disposable filter with 6,35 mm connection, 6,35 mm vent/drain compression seal fittings, Ø×L: 69×113 mm, 0,05 µm, 1800 cm²
Katalogové číslo ENTECWAZ0S1S3
Dodavatel: ENTEGRIS
Přejít na:
Product Details & Documents
Photochemical filter with superior contaminant retention and low hold-up volume performance in sub-10 to 180 nm technology nodes.
Specifikace
- Filtrační plocha:1800 cm²
- Velikost pórů:0,05 µm
- Type:D disposable filter with 6,35 mm connection, 6,35 mm vent/drain compression seal fittings, Ø×L: 69×113 mm
- Sekvence:07