Orders LinkContactUsLinkComponent
[EN]DISPERSING AGGREGATE PT-DA 25/2EC-E1 1 * 1 KS
[EN]DISPERSING AGGREGATE PT-DA 25/2EC-E1 1 * 1 KS
Katalogové číslo KINE11030264
Dodavatel:   KINEMATICA
[EN]DISPERSING AGGREGATE PT-DA 25/2EC-E1 1 * 1 KS
Katalogové číslo KINE11030264
Dodavatel:  KINEMATICA
[EN]DISPERSING AGGREGATE PT-DA 25/2EC-E1 1 * 1 KS
[EN]DISPERSING AGGREGATE PT-DA 25/2EC-E1 1 * 1 KS
Price
56 210,00 Kč