Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Přihlásit se
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CHROMIUM STANDARD FOR ICP 1000 MG/L CR I 1 * 100 mL
Catalog # SUPL74582-100ML
Supplier:
Merck
Expand image
CHROMIUM STANDARD FOR ICP 1000 MG/L CR I 1 * 100 mL
Catalog # SUPL74582-100ML
Supplier:
Merck
Frequently Bought Together
Standard solutions for analytical quality assurance, Supelco®
Supplier: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
Chlorid draselný 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Dusičnan 1.000 mg/l ve vodě (z NaNO<sub>3</sub>) Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
VWR® AVS TITRINORM, pufrovací roztoky, 20 °C
Supplier: VWR Chemicals
Roztoky standardní pro měření vodivosti
Supplier: VWR Chemicals