Micro protein quantification set, QuantiMate
004, Quantification set, 15 mm, Quartz
Katalogové číslo 730-1450
Dodavatel: BIOCHROM
Přejít na:
Product Details & Documents
QuantiMate is designed for concentration and purity determinations of nucleic acids and proteins.
- Measurement performance giving accurate and reproducible concentration results
- Samples measured without time consuming dilutions
- Only one cuvette is required
Dimensions are equivalent to standard cuvettes in order to work in most spectrophotometers.
Specifikace
- Materiál okna:Quartz
- Popis:Quantification set
- Výška:15 mm
- Sekvence:004