Orders LinkContactUsLinkComponent
Flame atomisation systems
Flame atomisation systems
001, Flame atomisation system kit
Catalog # VARI190034100
Supplier:   VARIAN
Flame atomisation systems
Catalog # VARI190034100
Supplier:  VARIAN
Flame atomisation systems
Flame atomisation systems
001, Flame atomisation system kit
Price
86 177,52 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
The Mark 7 atomisation system handles complex matrices and achieves precise results quickly.
  • Contoured design for outstanding resistance to blockage even with the most difficult examples
  • Corrosion-resistant components
  • For high-acid matrices
Mark 7 system includes removable twin-headed mixing paddles in order to offering thorough mixing, super-fine aerosol and for precise, accurate determinations.
This system provides the capability to introduce high matrix samples and the flexibility to achieve high sensitivity. In this system the operator can tune performances by the external adjustable impact beads.
Ordering information: An adjustable barrel nebuliser, nebuliser-cleaning wire, PTFE impact beads, a burner cleaning and alignment card, and high-vacuum capillary tubings are available separately.
Specifications
  • Popis:
    Flame atomisation system kit
  • Sekvence:
    001
Accessories
Product Information
  • Catalog #
    Sekvence
    Popis
    Availability
    Price
  • VARI6910009700
    907
    Nitrile O-ring for capillary assembly in spray chamber ; i.Ø = 1/32" o.Ø = 3/32"
    Log in for availability
    1 * 1 KS
Product Information
  • Catalog #
    Sekvence
    Popis
    Availability
    Price
  • VARI9910025700
    903
    Glass impact beads
    Log in for availability
    1 * 5 KS - 6 076,62 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Availability
    Price
  • VARI9910093400
    905
    Aqueous spray chamber o-ring
    Log in for availability
    1 * 1 KS - 2 588,19 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Availability
    Price
  • VARI9910093500
    906
    Organics o-ring
    Log in for availability
    1 * 1 KS - 34 863,84 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Availability
    Price
  • VARI9910093600
    904
    Fluorinated mixing paddles
    Log in for availability
    1 * 1 KS - 7 529,28 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Availability
    Price
  • VARI9910105800
    908
    Integrated liquid trap (fluorinated float and magnet)
    Log in for availability
    1 * 1 KS - 12 685,20 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Availability
    Price
  • VARI9910093000
    902
    Nebuliser capillary kit
    Log in for availability
    1 * 1 KS - 22 853,82 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Availability
    Price
  • VARI9910093100
    908
    Integrated nebuliser block
    Log in for availability
    1 * 1 KS - 40 674,48 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Availability
    Price
  • VARI190034100
    001
    Flame atomisation system kit
    Log in for availability
    1 * 1 KS - 86 177,52 Kč