Operating supplies
01, Operating supplies set, SIPS 10/20
Katalogové číslo VARI190025400
Dodavatel: VARIAN
Přejít na:
Product Details & Documents
The operating supplies set includes two 500 ml constant pressure bottle, a diluent bottle (1 L), a three-way SIPS tee piece assembly for SIPS-20, six SIPS pump tubings, a SIPS kit and ten pack SIPS pump bands.
Specifikace
- Model:SIPS 10/20
- Popis:Operating supplies set
- Sekvence:01