Orders LinkContactUsLinkComponent
Labels, biohazard
Labels, biohazard
4, Biohazard labels, PIC 312-Tri 100-B7525, Hliník, Černá na žluté, 100×87 mm, 1 label/card
Catalog # 116-0727
Supplier:   Brady
Labels, biohazard
Catalog # 116-0727
Supplier:  Brady
Labels, biohazard
Labels, biohazard
4, Biohazard labels, PIC 312-Tri 100-B7525, Hliník, Černá na žluté, 100×87 mm, 1 label/card
Price
207,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
All signs comply with current European legislation.
Ordering information: The polyester labels have a permanent acrylic adhesive.
Specifications
  • Barva:
    Černá na žluté
  • Popis:
    Biohazard labels
  • Rozměry:
    100×87 mm
  • Materiál:
    Hliník
  • Sekvence:
    4
  • Model:
    PIC 312-Tri 100-B7525
  • Balené:
    1 label/card