Wafer baskets
03, Wafer basket, A14, 24 wafers (15,88 - 31,8 mm)
Katalogové číslo ENTEA14-01S-0215
Dodavatel: ENTEGRIS
Přejít na:
Product Details & Documents
These wafer baskets made of PFA are designed for processing and handling substrates. They are compatible with tanks and handles.
Compatible tanks:
A14 baskets: E12
A15 baskets: E12, E14-10-01, E22-101
A26 baskets: E14-25-01
A47 baskets: E14-21-01, E14-25-01
Compatible handles:
A14 baskets: A01, A07, A029
A15 baskets: A01, PA01, A07, PA07, A029, PA029
A26 baskets: A01, A07, A029
A47 baskets: A01, A07, A029
Specifikace
- Určeno pro:24 wafers (15,88 - 31,8 mm)
- Popis:Wafer basket, A14
- Sekvence:03
Accessories
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- For A14 wafer basket