Disk filtrační anal.uhlíkový,185mm 1 * 50 KS
BINZ146194
Frequently Bought Together
Dialysis cassettes, Slide-A-Lyzer™ G2, Pierce™
: Thermo Fisher Scientific
Quantitative filter papers, ashless grades, grade 41, Whatman™
: Whatman products (Cytiva)
Capsule filters, Polycap™ SPF, Whatman™
: Whatman products (Cytiva)
Filtr ze skleněných mikrovláken kruhový MN GF-5, 4,7cm 1 * 100 KS
: MACHEREY-NAGEL
Mesh woven filters, nylon, Spectra Mesh®
: Spectrum Laboratories
Nutrient Pad Sets, NKS e.motion
: Sartorius
Glass microfibre filters without binder
: Sartorius
[EN]MICROKROS 26CM 100KD MPES 1MM DRY 1 * 1 KS
: Spectrum Laboratories
Membrane disc filters, wwPTFE (water wettable PTFE)
: Cytiva (Formerly Pall Lab)
Papír filtrační kruhový MN 606 1/4, skládaný, 18,5cm 1 * 100 KS
: MACHEREY-NAGEL
Accessories for vacuum filtration units
: Sartorius