Orders LinkContactUsLinkComponent
Jars with screw cap
Jars with screw cap
02, with polypropylene screw cap, labelled, 150 ml
Katalogové číslo 216-2047
Dodavatel:   Anicrin
Jars with screw cap
Katalogové číslo 216-2047
Dodavatel:  Anicrin
Jars with screw cap
Jars with screw cap
02, with polypropylene screw cap, labelled, 150 ml
Přejít na:
Price
2 360,00 Kč
Přihlásit se pro dostupnost
1 * 250 KS
Product Details & Documents
Slightly conical vessels, made of PP, translucent with red PE screw cap, for sampling, storage and transportation.
  • Twist cap for quick opening and closing
  • Very good temperature resistance
  • Very good chemical resistance
Caution: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904
Specifikace
  • Type:
    with polypropylene screw cap, labelled
  • Kapacita:
    150 ml
  • Sekvence:
    02