Orders LinkContactUsLinkComponent
Control Cables
Control Cables
07, Control cable to connect Com.G@te® ECS to cable end open, 3 m
Katalogové číslo HUBE9491
Dodavatel:   HUBER
Control Cables
Katalogové číslo HUBE9491
Dodavatel:  HUBER
Control Cables
Control Cables
07, Control cable to connect Com.G@te® ECS to cable end open, 3 m
Přejít na:
Price
2 230,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
A range of control cables is available for USB, RS232, RS485 or analogue interfaces (AIF). Cables are also available for external control signal (ECS), a potentialfree contact (POKO) or for an external float switch (LEVEL).
Specifikace
  • Popis:
    Control cable to connect Com.G@te® ECS to cable end open
  • Délka:
    3 m
  • Sekvence:
    07