MEMANTINE (HYDROCHLORIDE) 1 * 25 mg
Katalogové číslo CAYM14184-25
Dodavatel: Cayman Chemical
Frequently Bought Together
Isohexane (mixture of isomers), EMSURE® p.a., Supelco®
Dodavatel: Merck
Acetonitril, anhydrous (max. 0.001% H₂O) ≥99.8%
Dodavatel: VWR Chemicals
Etylenglykol ≥99%, Reagent Grade
Dodavatel: Honeywell Chemicals
Dimethylsulfoxid, anhydrous ≥99.8%
Dodavatel: Thermo Fisher Scientific
Butanon 99+% for spectroscopy
Dodavatel: Thermo Fisher Scientific
Cyclopentyl methyl ether (CPME) ≥99.9%
Dodavatel: Thermo Fisher Scientific
Hexan (směs izomerů) 99% ACS
Dodavatel: Thermo Fisher Scientific
Butanon, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Dodavatel: Avantor
N,N-Dimethylacetamid 99.8+% for HPLC
Dodavatel: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran, anhydrous stabilizovaný for grignard-reaction
Dodavatel: Thermo Fisher Scientific
4-(Methylamino)phenol sulphate 99%, extra pure
Dodavatel: Thermo Fisher Scientific
Methanol with 0,1% (v/v) formic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
Dodavatel: Honeywell Chemicals
USP SOLUTION METHYL ORANGE 1 * 100 mL
Dodavatel: REAGECON