Orders LinkContactUsLinkComponent
Flasks, round bottom, with three parallel necks
Flasks, round bottom, with three parallel necks
01, 25 ml, 14/23 NS, 14/23 NS
Katalogové číslo NEUB0760-0025-14
Flasks, round bottom, with three parallel necks
Katalogové číslo NEUB0760-0025-14
Flasks, round bottom, with three parallel necks
01, 25 ml, 14/23 NS, 14/23 NS
Přejít na:
Price
2 070,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With white, matt labelling area.
Specifikace
  • Kapacita:
    25 ml
  • Sekvence:
    01
  • Boční hrdla:
    14/23 NS
  • Centrální hrdlo:
    14/23 NS